Order Entry
Export
ContactUsLinkComponent
2-Methyl-4(5)-nitroimidazole
2-Methyl-4(5)-nitroimidazole
Catalog # EHERC15107000
Supplier:  EHRENSTORFER
CAS Number:  
2-Methyl-4(5)-nitroimidazole
Catalog # EHERC15107000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :127.10 g/mol
EINECS:211-790-3
CAS Number:696-23-1
Formula:C₄H₅N₃O
MDL Number:MFCD00005191