2-Hydroxycarbazole
Glass bottle for solids, 5 g
Catalog # MOLE45893442-5G
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Carbazol-2-ol
- CAS Number:86-79-3
- Formula:C₁₂H₉NO
- MDL Number:MFCD00004962
- Molecular Weight:183.21 g/mol
- EINECS:201-699-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



