Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-4-iodo-3-methylpyridine
2-Chloro-4-iodo-3-methylpyridine 95%
2-Chloro-4-iodo-3-methylpyridine 95%
Irritant
2-Chloro-4-iodo-3-methylpyridine 95%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
2-Chloro-4-iodo-3-methylpyridine 95%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
153034-88-9
:
Formula:
C₆H₅ClIN
MDL Number:
MFCD03095293
:
253.47 g/mol
Melting Pt:
106…107 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH50107
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation