Order Entry
Export
Orders LinkContactUsLinkComponent
2-Chloro-4-hydroxypyridine

2-Chloro-4-hydroxypyridine

Glass bottle for solids, 1 g
Catalog # COBBPY-7235-1G
Irritant
Supplier:   COMBI-BLOCKS
2-Chloro-4-hydroxypyridine
Catalog # COBBPY-7235-1G
Supplier:  COMBI-BLOCKS
CAS Number:  17368-12-6
2-Chloro-4-hydroxypyridine

Glass bottle for solids, 1 g

CAS Number:  17368-12-6
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:17368-12-6
  • Formula:C₅H₄ClNO
  • MDL Number:MFCD01646108
  • Molecular Weight:129.55 g/mol
  • EINECS:241-399-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications