Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pyridine-2,6-dicarbonitrile
Pyridine-2,6-dicarbonitrile, Sigma-Aldrich®
Pyridine-2,6-dicarbonitrile, Sigma-Aldrich®
Pyridine-2,6-dicarbonitrile, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Pyridine-2,6-dicarbonitrile, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
2893-33-6
:
Formula:
C₇H₃N₃
MDL Number:
MFCD00129021
:
129.12 g/mol
EINECS:
220-766-1