2,4-Dimethylbenzenesulphonyl chloride, Sigma-Aldrich®
Catalog # SIAL714453-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:204.68 g/mol
UN:3261
ADR:8,II
CAS Number:609-60-9
Formula:C₈H₉ClO₂S
MDL Number:MFCD00156143