1,1,1-Tris(hydroxymethyl)ethane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:77-85-0
:
Formula:C₅H₁₂O₃
MDL Number:MFCD00004687
:120.15 g/mol
Boiling Pt:287 °C (1013 hPa)
:M-Clarity™ quality level = MQ200
EINECS:201-063-9