2-Nitro-1-naphthol, Sigma-Aldrich®
Glass bottle for solids, 5 g
Catalog # SIAL161152-5G
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: 1-Hydroxy-2-nitronaphthalene
- CAS Number:607-24-9
- Formula:C₁₀H₇NO₃
- MDL Number:MFCD00003956
- Molecular Weight:189.17 g/mol
- Melting Pt:123…125 °C
- :M-Clarity™ quality level = MQ100
- EINECS:210-131-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



