2,4,5-Trichlorophenol
Glass bottle for solids, 100 mg
Catalog # ACCUA-025
Supplier: AccuStandard
Jump to:
Product Details & Documents
- CAS Number:95-95-4
- Formula:C₆H₃Cl₃O
- MDL Number:MFCD00002170
- Molecular Weight:197.45 g/mol
- Merck Index:12,09772
- Boiling Pt:253 °C (1013 hPa)
- Melting Pt:68 °C
- Density:1.68 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:202-467-8
- UN:3077
- ADR:9,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



