Pesticide reference standard, 2,3,4,6-Tetrachlorophenol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA17374600ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:58-90-2
- Description:2,3,4,6-Tetrachlorophenol 100 µg/ml in methanol
- Formula:C₆H₂Cl₄O
- Description:2,3,4,6-Tetrachlorophenol