Order Entry
Export
ContactUsLinkComponent
Organic reference standard, o-Ethyltoluene 100 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, o-Ethyltoluene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA13356200ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, o-Ethyltoluene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA13356200ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    611-14-3
  • Description:
    o-Ethyltoluene 100 µg/ml in methanol
  • Formula:
    C₉H₁₂
  • Description:
    o-Ethyltoluene

Specifications

About this item