Order Entry
Export
Orders LinkContactUsLinkComponent
 

4'-Hexylacetophenone

Supplier:   Thermo Scientific
4'-Hexylacetophenone
Supplier:  Thermo Scientific
Certificates
4'-Hexylacetophenone

3 Options Available Below

Product Details & Documents
Synonyms:  p-Hexylacetophenone, 1-(4-Hexylphenyl)ethanone
  • CAS Number:37592-72-6
  • Formula:C₁₄H₂₀O
  • MDL Number:MFCD00043677
  • Molecular Weight:204.31
Product Family Options

Product Information