

3,4-Dimethoxybenzonitrile ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2024-83-1
:
Formula:C₉H₉NO₂
MDL Number:MFCD00001802
:163.18 g/mol
Boiling Pt:131…133 °C (0.3 mmHg)
Melting Pt:65…69 °C
Storage Temperature:Ambient
EINECS:217-969-2
UN:3439
ADR:6.1,III
:GHS06
:GHS07