2-Chloro-5-(trifluoromethyl)phenol, Sigma-Aldrich®
Catalog # SIAL365807-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:196.56 g/mol
Boiling Pt:87…88 °C (38 mmHg)
Density:1.459 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
CAS Number:40889-91-6
Formula:C₇H₄ClF₃O
MDL Number:MFCD00019995