Order Entry
Export
Orders LinkContactUsLinkComponent
4-Bromobenzonitrile

4-Bromobenzonitrile

Supplier:   COMBI-BLOCKS
4-Bromobenzonitrile
Supplier:  COMBI-BLOCKS
Certificates
4-Bromobenzonitrile

2 Options Available Below

Product Details & Documents
Synonyms:  p-Bromobenzonitrile
  • CAS Number:623-00-7
  • Formula:C₇H₄BrN
  • MDL Number:MFCD00001811
  • Molecular Weight:182.02 g/mol
  • Boiling Pt:236 °C (1013 hPa)
  • Melting Pt:∼90 °C
  • Density:1.562 g/cm³ (20 °C)
  • Flash Pt:130 °C
  • Storage Temperature:Ambient
  • EINECS:210-764-9
  • UN:3439
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g