5-Bromo-2-chlorophenol
Supplier: COMBI-BLOCKS
Jump to:
About this item
- CAS Number:183802-98-4
- Formula:C₆H₄BrClO
- MDL Number:MFCD00672940
- Molecular Weight:207.45 g/mol
2 Options Available Below
Product Details & Documents
- CAS Number:183802-98-4
- Formula:C₆H₄BrClO
- MDL Number:MFCD00672940
- Molecular Weight:207.45 g/mol
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



