Order Entry
Export
Orders LinkContactUsLinkComponent
4-Bromo-2,6-dichlorophenol

4-Bromo-2,6-dichlorophenol

Glass bottle for solids, 5 g
Catalog # COBBOT-0804-5G
Supplier:   COMBI-BLOCKS
4-Bromo-2,6-dichlorophenol
Catalog # COBBOT-0804-5G
Supplier:  COMBI-BLOCKS
CAS Number:  3217-15-0
4-Bromo-2,6-dichlorophenol

Glass bottle for solids, 5 g

CAS Number:  3217-15-0
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:3217-15-0
  • Formula:C₆H₃BrCl₂O
  • MDL Number:MFCD00019996
  • Molecular Weight:241.9 g/mol
  • EINECS:221-740-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications