Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Bromo-3-chlorophenol

Glass bottle for solids, 25 g
Catalog # COBBAN-1044-25G
Supplier:   COMBI-BLOCKS
4-Bromo-3-chlorophenol
Catalog # COBBAN-1044-25G
Supplier:  COMBI-BLOCKS
CAS Number:  13631-21-5
4-Bromo-3-chlorophenol

Glass bottle for solids, 25 g

CAS Number:  13631-21-5
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:13631-21-5
  • Formula:C₆H₄BrClO
  • MDL Number:MFCD00070744
  • Molecular Weight:207.45 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications