Order Entry
Export
Orders LinkContactUsLinkComponent
4,4'-Diaminobenzophenone, Sigma-Aldrich®

4,4'-Diaminobenzophenone, Sigma-Aldrich®

Plastic bottle for solids, 5 g
Catalog # SIAL378259-5G
Supplier:   Merck
4,4'-Diaminobenzophenone, Sigma-Aldrich®
Catalog # SIAL378259-5G
Supplier:  Merck
CAS Number:  611-98-3
4,4'-Diaminobenzophenone, Sigma-Aldrich®

Plastic bottle for solids, 5 g

CAS Number:  611-98-3
Price
247,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Bis(4-aminophenyl)methanone, 4,4′-Diaminobenzophenone
  • CAS Number:611-98-3
  • Formula:C₁₃H₁₂N₂O
  • MDL Number:MFCD00038138
  • Molecular Weight:212.25 g/mol
  •  :M-Clarity™ quality level = MQ100
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 g

Specifications