Order Entry
Export
ContactUsLinkComponent
Pesticide reference standard, 3,4-Dichloroaniline 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Pesticide reference standard, 3,4-Dichloroaniline 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA12323400AL
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 3,4-Dichloroaniline 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA12323400AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    95-76-1
  • Description:
    3,4-Dichloroaniline 100 µg/ml in acetonitrile
  • Formula:
    C₆H₅Cl₂N
  • Description:
    3,4-Dichloroaniline

Specifications

About this item