Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-5-fluoroaniline
2-Chloro-5-fluoroaniline ≥97%
2-Chloro-5-fluoroaniline ≥97%
:
Thermo Fisher Scientific
2-Chloro-5-fluoroaniline ≥97%
:
Thermo Fisher Scientific
:
452-83-5
:
:
C₆H₅ClFN
:
MFCD00084852
:
145.56 g/mol
:
211 °C
:
∼ 26 °C
:
2811
:
6.1,III
:
GHS06
:
GHS08
:
GHS07
:
Ambient
Environmentally Preferable:
msds_ALFAB25618
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years