Dexamethasone-21-acetate
Catalog # PROOICRS0268
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:434.5 g/mol
EINECS:214-646-8
CAS Number:1177-87-3
Formula:C₂₄H₃₁FO₆
MDL Number:MFCD00027407