Hydrocortisone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:50-23-7
:
Formula:C₂₁H₃₀O₅
MDL Number:MFCD00011654
Boiling Pt:567
Melting Pt:202…204 °C
Density:1.4 g/cm³ (20 °C)
Molecular Weight:362.47 g/mol
:M-Clarity™ quality level = MQ200
EINECS:200-020-1
Specifications
- Environmentally Preferable: