Hydrocortisone
Glass bottle for solids, 100 mg
Catalog # PROOICRS0325
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
- CAS Number:50-23-7
- Formula:C₂₁H₃₀O₅
- MDL Number:MFCD00011654
- Molecular Weight:362.47 g/mol
- Boiling Pt:567 °C (1013 hPa)
- Melting Pt:202…204 °C
- Density:1.4 g/cm³ (20 °C)
- EINECS:200-020-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



