Order Entry
Export
Orders LinkContactUsLinkComponent
Hydrocortisone

Hydrocortisone

Glass bottle for solids, 1 g
Catalog # FITZ30-AC19
Hydrocortisone
Catalog # FITZ30-AC19
CAS Number:  50-23-7
Hydrocortisone

Glass bottle for solids, 1 g

CAS Number:  50-23-7
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:50-23-7
  • Formula:C₂₁H₃₀O₅
  • MDL Number:MFCD00011654
  • Molecular Weight:362.47 g/mol
  • Boiling Pt:567 °C (1013 hPa)
  • Melting Pt:202…204 °C
  • Density:1.4 g/cm³ (20 °C)
  • EINECS:200-020-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications