Order Entry
Export
Orders LinkContactUsLinkComponent
Hydrocortisone

Hydrocortisone

Glass bottle, 1 mL
Catalog # FITZ20-CG50
Hydrocortisone
Catalog # FITZ20-CG50
CAS Number:  50-23-7
Hydrocortisone

Glass bottle, 1 mL

CAS Number:  50-23-7
Price
Inquire For Price
Log in for availability
1 * 1 mL
Product Details & Documents
  • CAS Number:50-23-7
  • Formula:C₂₁H₃₀O₅
  • MDL Number:MFCD00011654
  • Molecular Weight:362.47 g/mol
  • Boiling Pt:567 °C (1013 hPa)
  • Melting Pt:202…204 °C
  • Density:1.4 g/cm³ (20 °C)
  • EINECS:200-020-1
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL

Specifications