1,4-Dibenzoylbenzene, Sigma-Aldrich®
Catalog # SIAL445932-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:286.33 g/mol
:M-Clarity™ quality level = MQ100
EINECS:221-155-2
CAS Number:3016-97-5
Formula:C₂₀H₁₄O₂
MDL Number:MFCD00014087