Order Entry
Export
ContactUsLinkComponent
Organic reference standard, 3,5-Dinitrotoluene 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Organic reference standard, 3,5-Dinitrotoluene 100 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERXA12786800AL
 :  
Organic reference standard, 3,5-Dinitrotoluene 100 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERXA12786800AL
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    618-85-9
  • Description:
    3,5-Dinitrotoluene 100 µg/ml in acetonitrile
  • Formula:
    C₇H₆N₂O₄
  • Description:
    3,5-Dinitrotoluene