Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Difluoro-4-nitrobenzene
1,3-Difluoro-4-nitrobenzene
1,3-Difluoro-4-nitrobenzene
:
Thermo Scientific
1,3-Difluoro-4-nitrobenzene
:
Thermo Scientific
:
446-35-5
:
:
C₆H₃F₂NO₂
:
MFCD00007050
:
159.09 g/mol
:
203 °C (1013 hPa)
:
10 °C
:
1.4571 g/cm³ (14 °C)
:
207-167-0
:
2810
:
6.1,III