3,5-Dibromophenol
Glass bottle for solids, 5 g
Catalog # COBBOT-0545-5G
Supplier: COMBI-BLOCKS
Jump to:
Product Details & Documents
- CAS Number:626-41-5
- Formula:C₆H₄Br₂O
- MDL Number:MFCD06411376
- Molecular Weight:251.91 g/mol
- Boiling Pt:274 °C (1013 hPa)
- Melting Pt:81 °C
- Density:2,1 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



