Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Iodo-α,α,α,α',α',α'-Hexafluoro-m-xylene
5-Iodo-α,α,α,α',α',α'-hexafluoro-m-xylene
5-Iodo-α,α,α,α',α',α'-hexafluoro-m-xylene
5-Iodo-α,α,α,α',α',α'-hexafluoro-m-xylene
Supplier:
Thermo Scientific
Certificates
Expand image
5-Iodo-α,α,α,α',α',α'-hexafluoro-m-xylene
Supplier:
Thermo Scientific
About this item
CAS Number:
328-73-4
:
Formula:
C₈H₃F₆I
MDL Number:
MFCD00040837
:
340.01 g/mol
Boiling Pt:
171…172 °C (1013 hPa)
Density:
1,91 g/cm³ (20 °C)