Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dibenzoylbenzene
1,3-Dibenzoylbenzene ≥98%
1,3-Dibenzoylbenzene ≥98%
1,3-Dibenzoylbenzene ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
1,3-Dibenzoylbenzene ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
3770-82-9
:
Formula:
C₂₀H₁₄O₂
MDL Number:
MFCD00014086
:
286.33 g/mol
Melting Pt:
98…99 °C
Storage Temperature:
Ambient
EINECS:
223-210-6
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAA12768
Application Notes
Organic Building Blocks and Intermediates
Documentation