Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Iodo-o-xylene
4-Iodo-o-xylene
4-Iodo-o-xylene
Irritant
4-Iodo-o-xylene
Supplier:
Thermo Scientific
Certificates
Expand image
Irritant
4-Iodo-o-xylene
Supplier:
Thermo Scientific
About this item
CAS Number:
31599-61-8
:
Formula:
C₈H₉I
MDL Number:
MFCD00040989
:
232.06 g/mol
Boiling Pt:
231…233 °C (1013 hPa)
Density:
1,633 g/cm³ (20 °C)
:
GHS07