Pesticide reference standard, 2,3-Dichlorophenol 10 µg/ml in methanol, Dr Ehrenstorfer
EHERL12450000ME
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:576-24-9
- Description:2,3-Dichlorophenol 10 µg/ml in methanol
- Formula:C₆H₄Cl₂O
- Description:2,3-Dichlorophenol