3,4-Diaminobenzophenone, Sigma-Aldrich®
Catalog # SIAL191760-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:212.25 g/mol
Boiling Pt:390 °C (1013 hPa)
Melting Pt:116…118 °C
EINECS:254-273-8
CAS Number:39070-63-8
Formula:C₁₃H₁₂N₂O
MDL Number:MFCD00007727