

4,4'-Bis(dimethylamino)benzophenone
Catalog # MOLEM73866402
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:268.36 g/mol
Merck Index:12,06265
Boiling Pt:428 °C (1013 hPa)
Melting Pt:172 °C
Flash Pt:220 °C
Storage Temperature:Ambient
EINECS:202-027-5
UN:2811
ADR:6.1,III
:GHS05
:GHS08
CAS Number:90-94-8
Formula:C₁₇H₂₀N₂O
MDL Number:MFCD00008312