Tris(perfluorobutyl)amine
Catalog # EHERC15987500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:671.1 g/mol
Boiling Pt:155 °C (1013 hPa)
Melting Pt:–55 °C
Density:1,883 g/cm³ (20 °C)
EINECS:206-223-1
CAS Number:311-89-7
Formula:C₁₂F₂₇N
MDL Number:MFCD00000436