Order Entry
Export
Orders LinkContactUsLinkComponent
1,4-Dithioerythritol

1,4-Dithioerythritol

Irritant
Supplier:   Roth Carl
1,4-Dithioerythritol
Supplier:  Roth Carl
Certificates
1,4-Dithioerythritol

2 Options Available Below

Product Details & Documents
Synonyms:  Cleland's Reagent, (R*,S*)-1,4-Dimercaptobutane-2,3-diol, DTE, Erythro-1,4-Dimercaptobutane-2,3-diol, trans-1,4-Dimercaptobutane-2,3-diol, Dithioerythritol, DL-Dithioerythritol
  • CAS Number:6892-68-8
  • Formula:C₄H₁₀O₂S₂
  • MDL Number:MFCD00063750
  • Molecular Weight:154.25 g/mol
  • Boiling Pt:286 °C (1013 hPa)
  • Melting Pt:82…83 °C
  • Storage Temperature:Ambient
  • EINECS:229-998-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 90,90 €
    90,90 €
    1 * 5 g
       90,90 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 327,00 €
    327,00 €
    1 * 25 g
       327,00 €