Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(1S)-(+)-Camphor-10-sulphonic acid
(1S)-(+)-Camphor-10-sulphonic acid
(1S)-(+)-Camphor-10-sulphonic acid
(1S)-(+)-Camphor-10-sulphonic acid
MOLEM28017028
:
Molekula
:
(1S)-(+)-Camphor-10-sulphonic acid
MOLEM28017028
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
232.3 g/mol
:
375 °C (1013 hPa)
:
375 °C (1013 hPa)
:
195 °C
:
221-554-1
:
3144-16-9
:
C₁₀H₁₆O₄S
:
MFCD00064157