

D-(+)-Camphor, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:464-49-3
:
Formula:C₁₀H₁₆O
MDL Number:MFCD00064149
:152.24 g/mol
Merck Index:13,01739
Boiling Pt:408 °C (1013 hPa)
Melting Pt:174…175 °C
Density:0.992 g/cm³ (20 °C)
EINECS:207-355-2
UN:2717
ADR:4.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:66 °C
:GHS02
:GHS07
Storage Temperature:Ambient