Order Entry
Export
Orders LinkContactUsLinkComponent
Acetaldehyde, Sigma-Aldrich®

Acetaldehyde, Sigma-Aldrich®

Flammable
Health
Irritant
Supplier:   Merck
Acetaldehyde, Sigma-Aldrich®
Supplier:  Merck
Certificates
Acetaldehyde, Sigma-Aldrich®
Acetaldehyde, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  Ethanal
  • CAS Number:75-07-0
  • Formula:CH₃CHO
  • MDL Number:MFCD00006991
  • Molecular Weight:44.05 g/mol
  • Merck Index:12,00037
  • Boiling Pt:20.1 °C (1013 hPa)
  • Melting Pt:–123 °C
  • Density:0.78 g/cm³ (20 °C)
  • Flash Pt:–27 °C
  • Storage Temperature:Refrigerator
  • UN:1089
  • ADR:3,I
  •  :M-Clarity™ quality level = MQ200
  • EINECS:200-836-8

Documents

Certificates
Search by Lot Number
Product Family Options

Product Information