Order Entry
Export
Orders LinkContactUsLinkComponent
Laurinaldehyde

Laurinaldehyde

Glass bottle, 50 mL
Catalog # ROTH3754.1
Supplier:   Roth Carl
Laurinaldehyde
Catalog # ROTH3754.1
Supplier:  Roth Carl
CAS Number:  112-54-9
Laurinaldehyde

Glass bottle, 50 mL

CAS Number:  112-54-9
Price
67,40 €
Log in for availability
1 * 50 mL
Product Details & Documents
Synonyms:  Lauric aldehyde, Dodecylaldehyde, Dodecanal
  • CAS Number:112-54-9
  • Formula:C₁₂H₂₄O
  • MDL Number:MFCD00007017
  • Molecular Weight:184.32 g/mol
  • Boiling Pt:242 °C (1013 hPa)
  • Melting Pt:44 °C
  • Density:0.83 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:203-983-6
  • UN:3082
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL

Specifications