

1-Bromo-2,2-dimethoxypropane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:126-38-5
:
Formula:BrH₂CC(OCH₃)₂CH₃
MDL Number:MFCD00000208
Boiling Pt:83…87 °C (80 mmHg)
Density:1.354
Flash Pt:40 °C (104 °F)
EINECS:204-784-7
UN:3271
ADR:3,III
:183.05 g/mol
:GHS02
:GHS07
Storage Temperature:Ambient