
1,2-Dibromotetrachloroethane 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:630-25-1
:
Formula:BrCl₂CCBrCl₂
MDL Number:MFCD00000780
:325.64 g/mol
Boiling Pt:222 °C (1013 hPa)
Melting Pt:220…222 °C
Density:2.71 g/cm³
Storage Temperature:Ambient
EINECS:211-136-7
:GHS07
Specification Test Results
- Appearance:White to beige Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- GC:≥96.0 %