2,2-Dibromopropane 98%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:594-16-1
- Formula:H₃CBr₂CCH₃
- MDL Number:MFCD00000119
- Molecular Weight:201.89 g/mol
- Boiling Pt:114 °C (740 mmHg)
- Melting Pt:–20 °C
- Density:1.78 g/cm³
- Flash Pt:>110 °C
- EINECS:209-828-9
2 Options Available Below
Product Details & Documents
- CAS Number:594-16-1
- Formula:H₃CBr₂CCH₃
- MDL Number:MFCD00000119
- Molecular Weight:201.89 g/mol
- Boiling Pt:114 °C (740 mmHg)
- Melting Pt:–20 °C
- Density:1.78 g/cm³
- Flash Pt:>110 °C
- EINECS:209-828-9
Documents
ACRO18606
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4950 to 1.4990 (20°C, 589 nm)
- GC:≥97.5 %
- Residue after ignition:≤0.1 %
- Specific optical rotation:-66° to -69° (20°C, 589 nm) (1 % in EtOH)



