1,9-Dibromononane ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 4549-33-1 Molecular Formula: C9H18Br2 Formula Weight: 286.06 Flash Point: 110°(230°F) Refractive Index: 1.4965 Boiling Point: 285-288° Beilstein Reference: 1737598 MDL No.: MFCD00000279
Synonyms: Nonamethylene dibromide
- CAS Number:4549-33-1
- Formula:BrH₂C(CH₂)₇CH₂Br
- MDL Number:MFCD00000279
- Molecular Weight:286.05 g/mol
- Boiling Pt:285…288 °C
- Melting Pt:–10 °C
- Density:1.410
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:224-913-0
3 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 4549-33-1 Molecular Formula: C9H18Br2 Formula Weight: 286.06 Flash Point: 110°(230°F) Refractive Index: 1.4965 Boiling Point: 285-288° Beilstein Reference: 1737598 MDL No.: MFCD00000279
Synonyms: Nonamethylene dibromide
- CAS Number:4549-33-1
- Formula:BrH₂C(CH₂)₇CH₂Br
- MDL Number:MFCD00000279
- Molecular Weight:286.05 g/mol
- Boiling Pt:285…288 °C
- Melting Pt:–10 °C
- Density:1.410
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:224-913-0



