Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,6-Dibromohexane 98%

Irritant
Environment
1,6-Dibromohexane 98%
Certificates
1,6-Dibromohexane 98%

3 Options Available Below

Product Details & Documents
Synonyms:  Hexamethylen dibromide
  • CAS Number:629-03-8
  • Formula:BrH₂C(CH₂)₄CH₂Br
  • MDL Number:MFCD00000272
  • Molecular Weight:243.97 g/mol
  • Boiling Pt:243 °C (760 mmHg)
  • Melting Pt:–2 °C
  • Density:1.58 g/cm³
  • Flash Pt:110 °C
  • Storage Temperature:Ambient
  • EINECS:211-067-2
  • UN:3082
  • ADR:9,III

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5056 to 1.5076 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Specific optical rotation:
    +29° to +33° (20°C, 589 nm) (c=1, toluene)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 38,50 €
    38,50 €
    1 * 100 mL
       38,50 €

  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 100,00 €
    100,00 €
    1 * 500 mL
       100,00 €

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 451,00 €
    451,00 €
    1 * 2,5 L
       451,00 €