
1,4-Dibromopentane ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:626-87-9
:
Formula:BrH₂C(CH₂)₂BrCHCH₃
MDL Number:MFCD00000153
:229.94 g/mol
Boiling Pt:102…103 °C (15 mmHg)
Melting Pt:–34 °C
Density:1.687
Flash Pt:>110 °C (230 °F)
EINECS:210-967-2
:GHS07
Storage Temperature:Ambient