Order Entry
Export
Orders LinkContactUsLinkComponent
1,2-Dibromoethane

1,2-Dibromoethane

Glass bottle, 1 L
Catalog # 24025.292
Toxic
Health
Environment
Supplier:   VWR Chemicals
1,2-Dibromoethane
Catalog # 24025.292
Supplier:  VWR Chemicals
CAS Number:  106-93-4
1,2-Dibromoethane
1,2-Dibromoethane

Glass bottle, 1 L

CAS Number:  106-93-4
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
  • CAS Number:106-93-4
  • Formula:BrH₂CCH₂Br
  • MDL Number:MFCD00000233
  • Molecular Weight:187.86 g/mol
  • Merck Index:13,03830
  • Boiling Pt:131.7 °C (1013 hPa)
  • Melting Pt:10 °C
  • Density:2.18 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:203-444-5
  • UN:1605
  • ADR:6.1,I

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications