

1,2-Dibromobutane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:533-98-2
:
Formula:BrH₂CBrCHCH₂CH₃
MDL Number:MFCD00000157
:215.92 g/mol
Merck Index:14,1566
Boiling Pt:166…167 °C
Melting Pt:–65 °C
Density:1.789
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:208-581-4
:GHS06
:GHS07